missing translation for 'onlineSavingsMsg'
Learn More

Oglemilast, MedChemExpress

Product Code. 16492457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492457 50 mg 50mg
16456348 10 mg 10mg
16428937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492457 Supplier Medchem Express Supplier No. HY15178
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oglemilast (GRC 3886) is a potent and orally active phosphodiesterase-4 (PDE4) inhibitor with an IC50 of 0.5 nM for PDE4D3. Oglemilast inhibits pulmonary cell infiltration, including eosinophilia and neutrophilia in vitro and in vivo. Oglemilast has the potential for inflammatory airway diseases.

Oglemilast (GRC 3886) is a potent and orally active phosphodiesterase-4 (PDE4) inhibitor with an IC50 of 0.5 nM for PDE4D3. Oglemilast inhibits pulmonary cell infiltration, including eosinophilia and neutrophilia in vitro and in vivo. Oglemilast has the potential for inflammatory airway diseases.

Specifications

Chemical Name or Material Oglemilast
CAS 778576-62-8
Color Khaki
Purity Grade Notes Research
Molecular Formula C20H13Cl2F2N3O5S
Quantity 50 mg
Synonym GRC 3886
Solubility Information DMSO : 4 mg/mL (7.75 mM; Need ultrasonic)
SMILES O=C(C1=C2C(OC3=CC=C(NS(=O)(C)=O)C=C23)=C(OC(F)F)C=C1)NC4=C(Cl)C=NC=C4Cl
Molecular Weight (g/mol) 516.3
Formula Weight 516.3
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.