missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGG1 Rabbit anti-Human, Mouse, Rat, Clone: 8V2Q9, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | OGG1 |
|---|---|
| Clone | 8V2Q9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348874
|
Novus Biologicals
NBP3-16526-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18350656
|
Novus Biologicals
NBP3-16526-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OGG1 Monoclonal antibody specifically detects OGG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| OGG1 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 8-hydroxyguanine DNA glycosylase, 8-oxoguanine DNA glycosylase, AP lyase, DNA-apurinic or apyrimidinic site lyase, HOGG1, MMH, MUTMOGH1HMMH, N-glycosylase/DNA lyase | |
| A synthetic peptide corresponding to a sequence within amino acids 246-345 of human OGG1 (O15527). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8V2Q9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, Chromatin Research, DNA Repair | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4968 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title