missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OGG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA116505
This item is not returnable.
View return policy
Description
Suggested positive control: Jurkat whole cell extracts.
8-hydroxyguanine, a form of oxidative DNA damage induced by free radicals, causes G:C to T:A transversion. In E. Coli, three DNA repair enzymes exist to prevent the mutagenic effects of 8-hydroxyguanine. One of these enzymes, MutM, was found to have a functional yeast (yOgg1) and human (hOgg1) homologue. hOgg1 proteins efficiently released the 8-hydroxyguanine opposite the pyrimidine from DNA and cleaved the AP site in a manner similar to bacterial and yeast enzymes. Genetic backgrounds in control of the repair of damaged DNA are involved in the susceptibility to cancer development. The hOgg1 gene has been mapped to region 3p26. 2, a region showing loss of heterozygosity (LOH) in a variety of cancers. In particular, 3p25-p26 is a common LOH region in lung cancer.
Specifications
| OGG1 | |
| Polyclonal | |
| Unconjugated | |
| OGG1 | |
| 8-hydroxyguanine DNA glycosylase; 8-oxoguanine DNA glycosylase; 8-oxoguanine DNA-glycosylase 1; 8-oxoguanine-DNA-glycosylase; AP lyase; DNA-(apurinic or apyrimidinic site) lyase; DNA-apurinic or apyrimidinic site lyase; HMMH; HOGG1; Mmh; MUTM; N-glycosylase/DNA lyase; OGG1; OGG1 type 1f; OGH1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100155036, 103345505, 18294, 4968, 81528 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O08760, O15527, O70249 | |
| OGG1 | |
| A peptide derived from human Ogg1 (within amino acids 1-100). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Monkey, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction