missing translation for 'onlineSavingsMsg'
Learn More

OGFOD2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18406086 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406086 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406086 Supplier Novus Biologicals Supplier No. NBP176533IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OGFOD2 Polyclonal antibody specifically detects OGFOD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen OGFOD2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 2-oxoglutarate and iron-dependent oxygenase domain containing 2, DKFZp686H15154, EC 1.14.11.-, FLJ13491, FLJ37501, MGC120434, MGC120436,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human OGFOD2. The immunogen is located within amino acids 140 - 190 of OGFOD2. Amino Acid Squence: EHFEQSDMPKGRPNTMN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79676
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.