missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OGFOD1 Monoclonal Antibody (OTI1D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525473
This item is not returnable.
View return policy
Description
OGFOD1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
OGFOD1, a member of the 2-OG-Fe(II) dioxygenase family, plays an important role in ischemic survival and stress. It is an oxidoreductase containing a Prolyl 4-hydroxylase alpha subunit. An OGFOD1 iron binding residue is required for ATPAF1 gene expression. As a novel stress granule component, OGFOD1 plays important proapoptotic roles in the regulation of translation and HRI-mediated phosphorylation of eIF2alpha in cells.
Specifications
| OGFOD1 | |
| Monoclonal | |
| 0.41 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N543 | |
| OGFOD1 | |
| Full length human recombinant protein of OGFOD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1D9 | |
| Unconjugated | |
| OGFOD1 | |
| 2-oxoglutarate and iron dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; 4930415J21Rik; AA387199; AA939912; AW061076; Kiaa1612; mKIAA1612; OGFOD1; Prolyl 3-hydroxylase OGFOD1; RGD1308848; Termination and polyadenylation 1 homolog; TPA1; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55239 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction