missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OGFOD1 Polyclonal antibody specifically detects OGFOD1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | OGFOD1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human OGFOD1. The immunogen is located within the first 50 amino acids of OGFOD1. Amino Acid Squence: ETLKKQVAEAWSRRTPFS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?