missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76532-0.025mg
This item is not returnable.
View return policy
Description
OGFOD1 Polyclonal specifically detects OGFOD1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Specifications
| OGFOD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- | |
| Rabbit | |
| 63 kDa | |
| 0.025 mg | |
| Lipid and Metabolism | |
| 55239 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| Q8N543 | |
| OGFOD1 | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human OGFOD1. The immunogen is located within the first 50 amino acids of OGFOD1. Amino Acid Squence: ETLKKQVAEAWSRRTPFS | |
| Affinity Purified | |
| RUO | |
| Primary | |
| OGFOD1 antibody is predicted to not cross-react with other TPA1 protein family members. At least two isoforms are known to exist; this antibody will only detect the larger isform. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction