missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGFOD1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18484AF532
This item is not returnable.
View return policy
Description
OGFOD1 Polyclonal antibody specifically detects OGFOD1 in Human samples. It is validated for Western Blot
Specifications
| OGFOD1 | |
| Polyclonal | |
| Western Blot | |
| 2-oxoglutarate and iron-dependent oxygenase domain containing 1,2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1, EC 1.14.11, FLJ10826, KIAA1612TPA1, termination and polyadenylation 1, homolog, Termination and polyadenylation 1 homolog, TPA1EC 1.14.11.- | |
| Recognizes region between residues Between between 1 and 50.. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 55239 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction