missing translation for 'onlineSavingsMsg'
Learn More

Ogerin, MedChemExpress

Product Code. 16494368
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494368 10 mg 10mg
16465276 100 mg 100mg
16447097 5 mg 5mg
16433056 25 mg 25mg
16430697 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494368 Supplier Medchem Express Supplier No. HY11027910mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ogerin is a selective GPR68 positive allosteric modulator, with a pEC50 of 6.83. Ogerin shows inverse agonist and antagonist activity (Ki, 220 nM) at A2A receptoor and weak antagonist activity (Ki, 736 nM) at 5-HT2B receptor. Ogerin blocks recall in fear conditioning in mice.

Ogerin is a selective GPR68 positive allosteric modulator, with a pEC50 of 6.83. Ogerin shows inverse agonist and antagonist activity (Ki, 220 nM) at A2A receptoor and weak antagonist activity (Ki, 736 nM) at 5-HT2B receptor. Ogerin blocks recall in fear conditioning in mice.

Specifications

Chemical Name or Material Ogerin
CAS 1309198-71-7
Color White
Purity Grade Notes Research
Molecular Formula C17H17N5O
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (813.40 mM; Need ultrasonic)
SMILES OCC1=C(C2=NC(N)=NC(NCC3=CC=CC=C3)=N2)C=CC=C1
Molecular Weight (g/mol) 307.35
Formula Weight 307.35
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.