missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ogerin
Description
Ogerin is a selective positive allosteric modulator (PAM) of GPR68 (pEC50 = 6.83), which exhibits selectivity for GPR68 over 58 other GPCRs. Ogerin attenuates contextual-based fear memory in mice.
Specifications
Specifications
| Chemical Name or Material | 2-[4-Amino-6-[(phenylmethyl)amino]-1,3,5-triazin-2-yl]benzenemethanol |
| CAS | 1309198-71-7 |
| Quantity | 50 mg |
| Target | Orphan 7-TM Receptor Agonists |
| Molecular Formula | C17H17N5O |
| Formula Weight | 307.35 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?