missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGDHL Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94830-0.1ml
This item is not returnable.
View return policy
Description
OGDHL Polyclonal antibody specifically detects OGDHL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| OGDHL | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| mitochondrial, OGDC-E1-like, oxoglutarate dehydrogenase-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human OGDHL (NP_060715.2). MSQLRLLPSRLGVQAARLLAAHDVPVFGWRSRSSGPPATFPSSKGGGGSSYMEEMYFAWLENPQSVHKSWDSFFREASEEAFSGSAQPRPPSVVHESRSAVSSRTKTSKL | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 55753 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction