missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15212-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is immunized with the C-terminal and can recognize the isoform 1 and 2 with the MW of 116 kDa.
This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitochondrial matrix and uses thiamine pyrophosphate as a cofactor. A congenital deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.Specifications
| OGDH | |
| Polyclonal | |
| Unconjugated | |
| OGDH | |
| AKGDH, E1k, OGDC, OGDC E1, OGDH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18293, 360975, 4967 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02218, Q5XI78, Q60597 | |
| Ogdh | |
| OGDH Fusion Protein Ag7368 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction