missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OGDH Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94086-0.1ml
This item is not returnable.
View return policy
Description
OGDH Polyclonal antibody specifically detects OGDH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| OGDH | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| 2-oxoglutarate dehydrogenase complex component E1, 2-oxoglutarate dehydrogenase, mitochondrial, AKGDH, alpha KGD, alpha-KGD, E1k, OGDC, oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide), oxoglutarate decarboxylase, oxoglutarate dehydrogenase (succinyl-transferring) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 148-427 of human OGDH (NP_001003941.1). DPLGILDADLDSSVPADIISSTDKLGFYGLDESDLDKVFHLPTTTFIGGQESALPLREIIRRLEMAYCQHIGVEFMFINDLEQCQWIRQKFETPGIMQFTNEEKRTLLARLVRSTRFEEFLQRKWSSEKRFGLEGCEVLIPALKTIIDKSSENGVDYVIMGMPHRGRLNVLANVIRKELEQIFCQFDSKLEAADEGSGDVKYHLGMYHRRINRVTDRNITLSLVANPSHLEAADPVVMGKTKAEQFYCGDTEGKKVRPRERRARQIVKAPCSSMEFRSPT | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 4967 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction