missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin N-Oxide Acetic Acid Salt (Mixture of Diastereomers), TRC

Product Code. 30137129
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137129 250 mg 250mg
30005613 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137129 Supplier Toronto Research Chemicals Supplier No. TRCO245755250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1216511-73-7
Molecular Formula C18 H20 F N3 O5 . C2 H4 O2
Quantity 250 mg
Synonym Ofloxacin Imp. F (EP), 4-[(RS)-6-Carboxy-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-10-yl]-1-methylpiperazine 1-Oxide Hydrochloride, Ofloxacin Imp. F (EP) as Hydrochloride, Ofloxacin N-Oxide Hydrochloride, Ofloxacin Impurity F as Hydrochloride
InChI Formula InChI=1S/C18H20FN3O5.C2H4O2/c1-10-9-27-17-14-11(16(23)12(18(24)25)8-21(10)14)7-13(19)15(17)20-3-5-22(2,26)6-4-20;1-2(3)4/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);1H3,(H,3,4)
SMILES CC1COc2c(N3CC[N+](C)([O-])CC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O.CC(=O)O
IUPAC Name acetic acid;7-fluoro-2-methyl-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5,7,9(13),11-tetraene-11-carboxylic acid
Molecular Weight (g/mol) 437.419
Formula Weight 437.16
Percent Purity >95%
Chemical Name or Material Ofloxacin N-Oxide Acetic Acid Salt (Mixture of Diastereomers)
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.