missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ofloxacin N-Oxide Acetic Acid Salt (Mixture of Diastereomers), TRC
Specifications
Specifications
| CAS | 2731368-77-5 |
| Molecular Formula | C18 H20 F N3 O5 . Cl H |
| Quantity | 25 mg |
| Synonym | 4-[(RS)-6-Carboxy-9-fluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-10-yl]-1-methylpiperazine 1-Oxide Hydrochloride, Ofloxacin Imp. F (EP) as Hydrochloride, Ofloxacin N-Oxide Hydrochloride, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-4-oxido-1-piperazinyl)-7-oxo-, hydrochloride (1:1) (ACI) |
| InChI Formula | InChI=1S/C18H20FN3O5.ClH/c1-10-9-27-17-14-11(16(23)12(18(24)25)8-21(10)14)7-13(19)15(17)20-3-5-22(2,26)6-4-20;/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);1H |
| SMILES | Cl.CC1COc2c(N3CC[N+](C)([O-])CC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O |
| IUPAC Name | 7-fluoro-2-methyl-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.05,13]trideca-5(13),6,8,11-tetraene-11-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 413.83 |
| Formula Weight | 413.1154 |
| Percent Purity | >95% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?