missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ofloxacin, 98%, Thermo Scientific Chemicals

Product Code. 15345038 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15345038

Brand: Thermo Scientific Chemicals 455670250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ofloxacin, CAS # 82419-36-1, is a fluoroquinolone antibiotic with broad antibacterial activity against gram-positive and -negative bacteria, as well as against mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ofloxacin is a synthetic fluoroquinolone antibiotic that is an analog of nalidixic acid
  • Ofloxacin can directly interact with pneumococcal autolysin (LytA) at its choline binding domain, thus inhibiting the ability of LytA to degrade the pneumococcal cell wall
  • Ofloxacin is racemic and its (S)-enantiomer is levofloxacin

Applications

  • Ofloxacin may be used in pharmacokinetic laboratory studies and high-throughput drug screens
  • The DNA and protein-binding capabilities of ofloxacin have been studied in the laboratory
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82419-36-1
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYSA-N
PubChem CID 4583
ChEBI CHEBI:7731
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Type Ofloxacin
Melting Point 280.0°C to 275.0°C
Infrared Spectrum Authentic
Loss on Drying 0.2% max.
Quantity 25 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 361.37
Percent Purity 98%
Chemical Name or Material Ofloxacin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.