missing translation for 'onlineSavingsMsg'
Learn More

OF-1, MedChemExpress

Product Code. 16441447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441447 50 mg 50mg
16417957 5 mg 5mg
16415238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16441447 Supplier Medchem Express Supplier No. HY12518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OF-1 is a potent pan-BRPF bromodomain (BRD) inhibitor, with IC50 values of 270 nM, 1.2 μM for TRIM24 and BRPF1B, respectively.

OF-1 is a potent pan-BRPF bromodomain (BRD) inhibitor, with IC50 values of 270 nM, 1.2 μM for TRIM24 and BRPF1B, respectively.

Specifications

Chemical Name or Material OF-1
CAS 919973-83-4
Color White
Purity Grade Notes Research
Molecular Formula C17H18BrN3O4S
Quantity 50 mg
Solubility Information DMSO : 13 mg/mL (29.52 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(Br)C=C1C)(NC2=C(OC)C=C(N3C)C(N(C)C3=O)=C2)=O
Molecular Weight (g/mol) 440.31
Formula Weight 440.31
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.