missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ OF 1
GREENER_CHOICE

Product Code. 18778216
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18778216 50 mg 50mg
18768476 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18778216 Supplier Tocris Bioscience™ Supplier No. 5289/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective BRPF1B and BRPF2 inhibitor

OF 1 is a selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.
TRUSTED_SUSTAINABILITY

Specifications

Purity 98%
Molecular Formula 4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide
CAS 919973-83-4
Content And Storage 4°C
Quantity 50 mg
Product Type OF 1
Molecular Weight (g/mol) M.W. (Observed): 440.31
Inhibitors OF 1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.