missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OF 1

Description
OF 1 is a selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.
Specifications
Specifications
| Purity | 98% |
| Molecular Formula | 4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide |
| CAS | 919973-83-4 |
| Content And Storage | 4°C |
| Quantity | 10 mg |
| Product Type | OF 1 |
| Molecular Weight (g/mol) | M.W. (Observed): 440.31 |
| Inhibitors | OF 1 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?