missing translation for 'onlineSavingsMsg'
Learn More

ODZ3 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18420067 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420067 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420067 Supplier Novus Biologicals Supplier No. NBP281974R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ODZ3 Polyclonal antibody specifically detects ODZ3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODZ3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias FLJ10474, FLJ10886, KIAA1455odd Oz/Ten-m homolog 3, odz, odd Oz/ten-m homolog 3 (Drosophila), ODZ3-like protein, Protein Odd Oz/ten-m homolog 3, ten-3, tenascin-M3, teneurin-3, Ten-m3, TNM3
Host Species Rabbit
Immunogen ODZ3 antibody was raised against a 17 amino acid peptide near the amino terminus of human ODZ3. The immunogen is located within the first 50 amino acids of ODZ3. Amino Acid Squence: SLTKSRREKERRYTNS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 55714
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.