Learn More
Odyssey™ Blocking Buffer (PBS), LI-COR Biosciences™
A non-mammalian protein blocker in phosphate-buffered saline. LI-COR Biosciences™ Odyssey™ Blocking Buffer (PBS) is suitable for use in a wide range of applications, including Western blots.
Brand:  Licor Biosciences 927-40000
Encompass
Product Code. 15590545
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
LI-COR offers blocking buffers for use with the Odyssey™ System. It is optimized for use with IRDye™ products and other near-infrared fluorophores, and provides excellent performance for immunoassays. Blocking buffers enhance sensitivity by reducing background interference, increasing signal-to-noise ratio, and promoting specific binding of the primary antibody while minimizing non-specific interactions.
  • Contains no mammalian proteins, for the lowest cross-reactivity with mammalian antibodies
  • Ready-to-use formulation in phosphate buffered saline (PBS)
  • Excellent choice for biotin-streptavidin detection
  • Suitable for a wide range of applications, including Western blots, In-Cell Western™ assays, and protein arrays
  • Also available in kits with Immobilon™-FL PVDF membranes