missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODF4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
ODF4 Polyclonal antibody specifically detects ODF4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ODF4 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | cancer/testis antigen 134, cancer/testis antigen 136, CT134, CT136, hOPPO1, MGC138215, OPPO1MGC138219, outer dense fiber 4, outer dense fiber of sperm tails 4, Outer dense fiber of sperm tails protein 4, outer dense fiber protein 4, Testis-specific protein oppo 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: FNHKSFWSLILSHPSGAVSCSSSFGSVEESPRAQTITDTPITQEGVLDPEQK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?