Learn More
Invitrogen™ ODF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139979
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FNHKSFWSLI LSHPSGAVSC SSSFGSVEES PRAQTITDTP ITQEGVLDPE QK Highest antigen sequence indentity to the following orthologs: Mouse - 35%, Rat - 37%.
ODF4 is the component of the outer dense fibers (ODF) of spermatozoa which could be involved in sperm tail structure, sperm movement and general organization of cellular cytoskeleton.This gene encodes a protein that is localized in the outer dense fibers of the tails of mature sperm. This protein is thought to have some important role in the sperm tail. Sequence Note: The RefSeq record was created from transcript and genomic sequence data to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on transcript alignments.
Specifications
| ODF4 | |
| Polyclonal | |
| Unconjugated | |
| ODF4 | |
| cancer/testis antigen 134; cancer/testis antigen 136; CT134; CT136; hOPPO1; MGC138215; MGC138219; ODF4; OPPO 1; OPPO1; outer dense fiber 4; outer dense fiber of sperm tails 4; outer dense fiber of sperm tails protein 4; Outer dense fiber protein 4; testis-specific protein oppo 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 146852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2M2E3 | |
| ODF4 | |
| Recombinant protein corresponding to Human ODF4. Recombinant protein control fragment (Product #RP-109280). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.