missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ODF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558709
This item is not returnable.
View return policy
Description
Immunogen sequence: PGPKYLIPPT TGFMKHTPTK LRAPAYSFRG APMLLAENCS PGPRYNVNPK ILRTGKDLGP AYSILGRYQT KTMLTP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
Outer dense fibers are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail. May help to maintain the passive elastic structures and elastic recoil of the sperm tail.
Specifications
| ODF3 | |
| Polyclonal | |
| Unconjugated | |
| Odf3 | |
| 1700011O04Rik; Beta390; cancer/testis antigen 135; CT135; Odf3; outer dense fiber of sperm tails 3; outer dense fiber of sperm tails protein 3; Outer dense fiber protein 3; SHIPPO1; Sperm tail protein SHIPPO 1; sperm tail protein SHIPPO1; TISP50; Transcript induced in spermiogenesis protein 50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 113746 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PU9 | |
| Odf3 | |
| Recombinant protein corresponding to Human ODF3. Recombinant protein control fragment (Product #RP-97513). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction