missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12058-1-AP-150UL
This item is not returnable.
View return policy
Description
Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintece of normal CETN1 assembly.Specifications
| ODF2 | |
| Polyclonal | |
| Unconjugated | |
| ODF2 | |
| Cenexin, ODF2, ODF2/1, ODF2/2, ODF84, Outer dense fiber protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18286, 29479, 4957 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A3KGV1, Q5BJF6, Q6AYX5 | |
| Odf2 | |
| ODF2 Fusion Protein Ag2693 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction