missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ODF2 Antibody (SR2297), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | ODF2 |
|---|---|
| Clone | SR2297 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:100 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601295
|
Novus Biologicals
NBP3-22037-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628634
|
Novus Biologicals
NBP3-22037-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ODF2 Monoclonal antibody specifically detects ODF2 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| ODF2 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cenexin, FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF8484-kD, outer dense fiber of sperm tails 2, outer dense fiber protein 2, sperm tail structural protein | |
| A synthesized peptide derived from human ODF2 (Uniprot #: Q5BJF6) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2297 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4957 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title