missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ODF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-85417
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
ODF2 Polyclonal specifically detects ODF2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Tekniske data
| ODF2 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Cenexin, FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF8484-kD, outer dense fiber of sperm tails 2, outer dense fiber protein 2, sperm tail structural protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4957 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ODF2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:HKRGMKGDTVNVRRSVRVKTKNPPHCLEITPPSSEKLVSVMRLSDLSTEDDDSGHCKMNRYDKKIDSLMNAVGCLKSEVKMQKGERQMAKRFLEERKEELEEVAHELAETEHENTVLRHNIERMKEEKDFTILQKKHLQQEKE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion