missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ OD 36 hydrochloride
Description
OD 36 hydrochloride is a potent receptor-interacting protein kinase 2 (RIPK2) inhibitor (IC50 = 5.3 nM). Displays similar % inhibition against ALK-2 and SIK2 at 100 nM. Downregulates expression of a panel of RIPK2-dependent genes. Inhibits recruitment of inflammatory cells in a mouse MDP-induced peritonitis model.
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-10,11,14,17-tetrahydro-13H-1,16-etheno-4,8-metheno-1H-pyrazolo[3,4-g][1,14,4,6]dioxadiazacyclohexadecine hydrochloride |
| CAS | 1638644-62-8 |
| Quantity | 10 mg |
| Molecular Formula | C16H15ClN4O2.HCl |
| Formula Weight | 367.23 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?