missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ocular development associated gene Antibody (OTI1H1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73142PCP
This item is not returnable.
View return policy
Description
Ocular development associated gene Monoclonal antibody specifically detects Ocular development associated gene in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Ocular development associated gene | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1H1 | |
| Western Blot | |
| FLJ22489, GATA zinc finger domain containing 1, GATA zinc finger domain-containing protein 1, ocular development associated, Ocular development-associated gene protein, ODAGFLJ40695, RG083M05.2 | |
| Human recombinant protein fragment corresponding to amino acids 68-269 of human GATAD1 (NP_066990) produced in E.coli. | |
| 0.1 mL | |
| Vision | |
| 57798 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction