missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ocular development associated gene Antibody (OTI1H1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73142AF405
This item is not returnable.
View return policy
Description
Ocular development associated gene Monoclonal specifically detects Ocular development associated gene in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Ocular development associated gene | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GATAD1 | |
| Recombinant protein fragment corresponding to amino acids 68-269 of human Ocular development associated gene (NP_066990) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1H1 | |
| Western Blot | |
| FLJ22489, GATA zinc finger domain containing 1, GATA zinc finger domain-containing protein 1, ocular development associated, Ocular development-associated gene protein, ODAGFLJ40695, RG083M05.2 | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 57798.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction