missing translation for 'onlineSavingsMsg'
Learn More

Octylmethyldichlorosilane, 97%, Thermo Scientific™

Product Code. 10663753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10663753 5 mL Glass bottle 5mL
10247590 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10663753 Supplier Thermo Scientific Acros Supplier No. 255260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14799-93-0
Molecular Formula C9H20Cl2Si
Molecular Weight (g/mol) 227.24
MDL Number MFCD00042606
InChI Key QHBMMABVNRSRHW-UHFFFAOYSA-N
Synonym n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
PubChem CID 84649
IUPAC Name dichloro-methyl-octylsilane
SMILES CCCCCCCC[Si](C)(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 230°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C8H17Si(CH3)Cl2
Refractive Index 1.442
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: .. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic(chlorinated) hydrocarbons
Formula Weight 227.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Octylmethyldichlorosilane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Octylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.