missing translation for 'onlineSavingsMsg'
Learn More

Octylmagnesium chloride, 1.4M solution in THF, AcroSeal™

Product Code. 10001084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001084 800 mL AcroSeal™ Glass bottle 800mL
10274824 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001084 Supplier Thermo Scientific Acros Supplier No. 428038000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 38841-98-4 | C8H17ClMg | 172.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38841-98-4 , 109-99-9
Molecular Formula C8H17ClMg
Molecular Weight (g/mol) 172.98
MDL Number MFCD00000476
InChI Key DPBWWQDKVQLWFK-UHFFFAOYSA-M
Synonym octylmagnesium chloride, magnesium, chlorooctyl, n-octylmagnesiumchloride, octyl magnesium chloride, n-c8h17mgcl, n-octylmagnesium chloride, octylmagnesium chloride solution, dpbwwqdkvqlwfk-uhfffaoysa-m, 1-octylmagnesium chloride, 1m in methf, octylmagnesium chloride solution, 2.0 m in thf
PubChem CID 4147387
SMILES CCCCCCCC[Mg]Cl

Specifications

CAS Min % 22.34, 70.21
CAS Max % 29.79, 77.66
Quantity 800 mL
Linear Formula CH3(CH2)6CH2MgCl
Flash Point −10°C
IUPAC Name chloro(octyl)magnesium
Formula Weight 172.98
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.929
Density 0.9290g/mL
Chemical Name or Material Octylmagnesium chloride, 1.4M solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Octylmagnesium chloride, 1.4M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.