missing translation for 'onlineSavingsMsg'
Learn More

Octyl chloroformate, 97%, Thermo Scientific Chemicals

Product Code. 10658634
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7452-59-7
Molecular Formula C9H17ClO2
Molecular Weight (g/mol) 192.69
MDL Number MFCD00000652
InChI Key VFXVAXFIFHSGNR-UHFFFAOYSA-N
Synonym octyl chloroformate, n-octyl chloroformate, chloroformic acid n-octyl ester, carbonochloridic acid, octyl ester, chloroformic acid octyl ester, n-octylchloroformate, octyl chlorocarboxylate, octyl chloridocarbonate #, acmc-209ov2
PubChem CID 81956
IUPAC Name octyl carbonochloridate
SMILES CCCCCCCCOC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9840g/mL
Boiling Point 90.0°C to 91.0°C (11.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClCO2(CH2)7CH3
Refractive Index 1.4300 to 1.4320
Quantity 25 g
Specific Gravity 0.984
Solubility Information Solubility in water: slightly soluble.
Formula Weight 192.69
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Octyl chloroformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'