missing translation for 'onlineSavingsMsg'
Learn More

Octyl aldehyde, 99%

Product Code. 10285220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285220 100 mL Glass bottle 100mL
10725231 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285220 Supplier Thermo Scientific Acros Supplier No. 129481000
  • ONLINE EXCLUSIVE PRICE
    £20.60
    £18.65 / 100mL
    Save £1.95
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124-13-0 | C8H16O | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-13-0
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00007029
InChI Key NUJGJRNETVAIRJ-UHFFFAOYSA-N
Synonym caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
PubChem CID 454
ChEBI CHEBI:17935
IUPAC Name octanal
SMILES CCCCCCCC=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 12.0°C to 15.0°C
Color Colorless to Yellow
Density 0.8210g/mL
Boiling Point 171.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)6CHO
Refractive Index 1.4170 to 1.4200
Quantity 100 mL
Beilstein 01, 704
Merck Index 15, 1768
Specific Gravity 0.821
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and mineral oil, insoluble in glycerol
Formula Weight 128.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Octyl aldehyde, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Octyl aldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.