missing translation for 'onlineSavingsMsg'
Learn More

Octopamine hydrochloride, MedChemExpress

Product Code. 16460978
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460978 5 g 5g
16442219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460978 Supplier Medchem Express Supplier No. HYB0528A
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Octopamine ((±)-p-Octopamine) hydrochloride, a biogenic monoamine structurally related to noradrenaline, acts as a neurohormone, a neuromodulator and a neurotransmitter in invertebrates.

Octopamine hydrochloride can stimulate alpha2-adrenoceptors (ARs) in Chinese hamster ovary cells transfected with human alpha2-ARs. Octopamine hydrochloride increased glycogenolysis, glycolysis, oxygen uptake, gluconeogenesis and the portal perfusion pressure.

Specifications

Chemical Name or Material Octopamine hydrochloride
CAS 770-05-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H12ClNO2
Quantity 5 g
Synonym (±)-p-Octopamine hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (527.31 mM) ∣H2O : ≥ 50 mg/mL (263.66 mM)
SMILES OC(C1=CC=C(O)C=C1)CN.Cl
Molecular Weight (g/mol) 189.64
Formula Weight 189.64
Percent Purity 99.28%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.