missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Octopamine hydrochloride, 99%

Código de producto. 15423899
Cambiar vista
Click to view available options
Quantity:
1 g
5 g
10 g
Tamaño de la unidad:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
15423899 1 g 1g
15433899 5 g 5g
15443899 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15423899 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J61281.03
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

alpha-adrenoceptor agonist; invertebrate neurotransmitter and neuromodulator | CAS: 770-05-8 | C8H12ClNO2 | 189.64 g/mol

Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octopamine HCL has the ability to halt catabolism of proteinaceous tissues. It is very important while losing weight to maximize adipose loss while minimizing the loss of muscle and other lean body tissue. Octopamine Hcl (norepinephrine) is best known as an effective ephedrine alternative, claiming to boost energy while reduce weight in bodybuilding industry for weight loss purpose. In addition, many users believe octopamine hcl is a good nootropic compound that makes our brain easy to focus, enhances motivation as a wakefulness promoter.

Solubility
It is soluble in water.

Notes
Handle under argon. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 770-05-8
Molecular Formula C8H12ClNO2
Molecular Weight (g/mol) 189.64
MDL Number MFCD00012881
InChI Key PUMZXCBVHLCWQG-UHFFFAOYNA-N
Synonym octopamine hydrochloride, dl-octopamine hydrochloride, octopamine hcl, epirenor, +/--octopamine hydrochloride, +,--octopamine hcl, 4-2-amino-1-hydroxyethyl phenol hydrochloride, +,--octopamine hydrochloride, 4-2-amino-1-hydroxyethyl phenol;hydrochloride
PubChem CID 102484
IUPAC Name 4-(2-amino-1-hydroxyethyl)phenol hydrochloride
SMILES Cl.NCC(O)C1=CC=C(O)C=C1

Especificaciones

Melting Point 170°C (decomposition)
Color White
Quantity 1 g
Beilstein 3915414
Merck Index 14,6759
Solubility Information It is soluble in water.
Formula Weight 189.64
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (+/-)-Octopamine hydrochloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.