missing translation for 'onlineSavingsMsg'
Learn More

Octopamine-13C2,15N Acetic Acid, TRC

Product Code. 30009811
Change view
Click to view available options
Quantity:
0.1 mg
1 mg
Unit Size:
0.1mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009811 1 mg 1mg
30007338 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009811 Supplier Toronto Research Chemicals Supplier No. TRCO2397541MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C2 C6 H11 15N O2 . C2 H4 O2
Molecular Weight (g/mol) 216.209
Synonym α-(Aminomethyl)-4-hydroxybenzenemethanol-13C2,15N Acetic Acid, (+/-)-Octopamine-13C2,15N Acetic Acid, (+/-)-p-Octopamaine-13C2,15N Acetic Acid, DL-Octopamine-13C2,15N Acetic Acid, Epirenor-13C2,15N Acetic Acid, NSC 108685-13C2,15N Acetic Acid, Norfen-13C2,15N Acetic Acid, Octopamine-13C2,15N Acetic Acid, dl-Octopamine-13C2,15N Acetic Acid
IUPAC Name acetic acid;4-(2-(^{15}N)azanyl-1-hydroxy-(1,2-^{13}C_{2})ethyl)phenol
SMILES CC(=O)O.[15NH2][13CH2][13CH](O)c1ccc(O)cc1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H11NO2.C2H4O2/c9-5-8(11)6-1-3-7(10)4-2-6;1-2(3)4/h1-4,8,10-11H,5,9H2;1H3,(H,3,4)/i5+1,8+1,9+1;
Formula Weight 216.1039
Chemical Name or Material Octopamine-13C2,15N Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.