missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octisalate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Octisalate
Specifications
Specifications
| Chemical Name or Material | Octisalate |
| CAS | 118-60-5 |
| Analyte or Component Names | Octisalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H22 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Hydroxybenzoic acid 2-ethylhexyl ester, 2-Ethylhexyl o-hydroxybenzoate, 2-Ethylhexyl salicylate, Dermoblock OS, Escalol 587, Eusolex OS, HallBrite OS-USP, NSC 46151, Neo Heliopan OS, Octisalate, Octyl salicylate, Parsol EHS, Salicylic acid, 2-ethylhexyl ester, Solarom OS, Sunarome O, Sunarome WMO, Uvinul 0-18, WMO, Salicylic acid-2-ethyl-1-hexyl ester |
| InChI Formula | InChI=1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3 |
| SMILES | CCCCC(CC)COC(=O)c1ccccc1O |
| IUPAC Name | 2-ethylhexyl 2-hydroxybenzoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction