missing translation for 'onlineSavingsMsg'
Learn More

Octisalate, MedChemExpress

Product Code. 16498516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498516 Supplier Medchem Express Supplier No. HYB0929
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Octisalate is an organic compound used as an ingredient in sunscreens and cosmetics to absorb the full range of UVB rays from the sun.

Octisalate is an organic compound used as an ingredient in sunscreens and cosmetics to absorb the full range of UVB rays from the sun.

Specifications

Chemical Name or Material Octisalate
CAS 118-60-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C15H22O3
Quantity 100 mg
Synonym Octyl salicylate 2-Ethylhexyl salicylate
Solubility Information DMSO : 100 mg/mL (399.47 mM; Need ultrasonic)
SMILES O=C(OCC(CC)CCCC)C1=CC=CC=C1O
Molecular Weight (g/mol) 250.33
Formula Weight 250.33
Percent Purity 98.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.