missing translation for 'onlineSavingsMsg'
Learn More

Octinoxate, Mikromol™

Product Code. 17979094
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17979094 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17979094 Supplier Mikromol Supplier No. MM0729.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Octinoxate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octinoxate
CAS 5466-77-3
Analyte or Component Names Octinoxate
Shipping Condition Room Temperature
Molecular Formula C18 H26 O3
Certifications/Compliance ISO 17034
Synonym Octinoxate, 2-Ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate, 2-Ethylhexyl 4-methoxycinnamate, 2-Ethylhexyl methoxycinnamate, 2-Ethylhexyl p-methoxycinnamate, 3-(4-Methoxyphenyl)-2-propenoic acid 2-ethylhexyl ester, 4-Methoxycinnamic acid 2-ethylhexyl ester, Escalol 557, Escalol 557NB, Escalol 557T, Ethylhexyl methoxycinnamate, Ethylhexyl p-methoxycinnamate, Eusolex 2292, Eusolex UV-Pearls OMC, Jeescreen OMC, NSC 26466, Neo Heliopan AV, Nomcort TAB, Nomcort TAB-R, Octyl 4-methoxycinnamate, Octyl methoxycinnamate, Octyl p-methoxycinnamate, Parsol MCX, Parsol MCX-SA, Parsol MOX, Solarom OMC, Sun Caps 664, Sunscreen AV, Tinosorb OMC, Uvinul 3088, Uvinul MC 80, Uvinul MC 80N, Uvinul MC 90, p-Methoxycinnamic acid 2-ethylhexyl ester
InChI Formula InChI=1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3/b13-10+
SMILES CCCCC(CC)COC(=O)\C=C\c1ccc(OC)cc1
IUPAC Name 2-ethylhexyl (E)-3-(4-methoxyphenyl)prop-2-enoate
Molecular Weight (g/mol) 290.4
Quantity 250 mg
Formula Weight 290.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.