missing translation for 'onlineSavingsMsg'
Learn More

Octinoxate, MedChemExpress

Product Code. 16482319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482319 Supplier Medchem Express Supplier No. HYB1234
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Octinoxate is an organic compound that is an ingredient in some sunscreens and lip balms, primarily used is in sunscreens and other cosmetics to absorb UV-B rays from the sun, protecting the skin from damage.

Octinoxate is an organic compound that is an ingredient in some sunscreens and lip balms, primarily used is in sunscreens and other cosmetics to absorb UV-B rays from the sun, protecting the skin from damage.

Specifications

Chemical Name or Material Octinoxate
CAS 5466-77-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H26O3
Quantity 1 g
Synonym Octyl methoxycinnamate
Solubility Information Ethanol : 140 mg/mL (482.09 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (3.44 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.31 mM; Need ultrasonic)
SMILES O=C(OCC(CC)CCCC)/C=C/C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 290.4
Formula Weight 290.4
Percent Purity 98.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.