missing translation for 'onlineSavingsMsg'
Learn More

Octenidine dihydrochloride, MedChemExpress

Product Code. p-200100469
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
5 g
Unit Size:
1g
200mg
5g
This item is not returnable. View return policy

Product Code. 16491466

Brand: Medchem Express HYB2170A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds.

Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds.

Specifications

Chemical Name or Material Octenidine dihydrochloride
CAS 70775-75-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H64Cl2N4
Quantity 200 mg
Solubility Information H2O : 33.33 mg/mL (53.43 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.02 mM; Need ultrasonic)
SMILES CCCCCCCC/N=C1C=CN(CCCCCCCCCCN(C=C/2)C=CC2=N/CCCCCCCC)C=C/1.Cl.Cl
Molecular Weight (g/mol) 623.83
Formula Weight 623.83
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.