missing translation for 'onlineSavingsMsg'
Learn More

Octenidine dihydrochloride, MedChemExpress

Product Code. 16491466
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
Unit Size:
1g
200mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491466 200 mg 200mg
16481148 5 g 5g
16462379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491466 Supplier Medchem Express Supplier No. HYB2170A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds.

Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds.

Specifications

Chemical Name or Material Octenidine dihydrochloride
CAS 70775-75-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H64Cl2N4
Quantity 200 mg
Solubility Information H2O : 33.33 mg/mL (53.43 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.02 mM; Need ultrasonic)
SMILES CCCCCCCC/N=C1C=CN(CCCCCCCCCCN(C=C/2)C=CC2=N/CCCCCCCC)C=C/1.Cl.Cl
Molecular Weight (g/mol) 623.83
Formula Weight 623.83
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.