missing translation for 'onlineSavingsMsg'
Learn More

Octaphenylcyclotetrasiloxane, 98+%

Product Code. 11332396
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332396 100 g 100g
11322396 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332396 Supplier Thermo Scientific Alfa Aesar Supplier No. L06130.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 546-56-5 | C48H40O4Si4 | 793.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-56-5
Molecular Formula C48H40O4Si4
Molecular Weight (g/mol) 793.18
MDL Number MFCD00003268
InChI Key VSIKJPJINIDELZ-UHFFFAOYSA-N
Synonym octaphenylcyclotetrasiloxane, cyclotetrasiloxane, octaphenyl, 2,2,4,4,6,6,8,8-octaphenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, octaphenyltetracyclosiloxane, cyclotetrasiloxane, 2,2,4,4,6,6,8,8-octaphenyl, 1,1,3,3,5,5,7,7-octaphenylcyclotetrasiloxane, octaphenyl tetracyclosiloxane, acmc-1aqwg, ph2sio 4, vsikjpjinidelz-uhfffaoysa
PubChem CID 68347
IUPAC Name 2,2,4,4,6,6,8,8-octakis-phenyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES O1[Si](O[Si](O[Si](O[Si]1(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 332°C (1mmHg)
Melting Point 193°C to 200°C
Quantity 100 g
Beilstein 2320758
Formula Weight 793.19
Percent Purity ≥98%
Chemical Name or Material Octaphenylcyclotetrasiloxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.