missing translation for 'onlineSavingsMsg'
Learn More

Octanoyl chloride, 99%

Product Code. 11372196
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372196 100 mL 100mL
11382196 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11372196 Supplier Thermo Scientific Alfa Aesar Supplier No. L03526.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 111-64-8 | C8H15ClO | 162.657 g/mol

Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-64-8
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.657
MDL Number MFCD00000766
InChI Key REEZZSHJLXOIHL-UHFFFAOYSA-N
Synonym capryloyl chloride, caprylyl chloride, n-octanoyl chloride, caprylic acid chloride, octanoic acid chloride, octanoic chloride, octanoylchloride, ccris 5990, caprylic chloride, octanoyl-chloride
PubChem CID 8124
IUPAC Name octanoyl chloride
SMILES CCCCCCCC(=O)Cl

Specifications

Melting Point -63°C
Density 0.942
Boiling Point 195°C
Flash Point 80°C (176°F)
Odor Pungent
Refractive Index 1.435
Quantity 100 mL
UN Number UN3265
Beilstein 635917
Sensitivity Moisture sensitive
Solubility Information Soluble in organic solvents (acetone,chloroform,toluene,THF). Hydrolyzes in water.
Formula Weight 162.66
Percent Purity 99%
Chemical Name or Material Octanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.