missing translation for 'onlineSavingsMsg'
Learn More

Octane, TRC

Product Code. 30422211
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity unitSize
30422211 1 L 1 L
1 options
This item is not returnable. View return policy

Product Code. 30422211

Brand: Toronto Research Chemicals TRCO2373701L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Octane
CAS 111-65-9
Molecular Formula C8 H18
Quantity 1 L
Synonym Octane, NSC 9822, n-Octane
InChI Formula InChI=1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3
SMILES CCCCCCCC
IUPAC Name octane
Molecular Weight (g/mol) 114.23
Formula Weight 114.1409
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.