missing translation for 'onlineSavingsMsg'
Learn More

Octanal, TRC

Product Code. 30425858
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425858 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425858 Supplier Toronto Research Chemicals Supplier No. TRCO23720050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octanal
CAS 124-13-0
Molecular Formula C8 H16 O
Quantity 50 g
Synonym Octanal, Aldehyde C 8, Antifoam LF, Caprylaldehyde, Caprylic aldehyde, NSC 1508, NSC 8969, Octaldehyde, Octanaldehyde, Octanoic aldehyde, Octylaldehyde, n-Caprylaldehyde, n-Octaldehyde, n-Octanal, n-Octyl aldehyde, n-Octylal
InChI Formula InChI=1S/C8H16O/c1-2-3-4-5-6-7-8-9/h8H,2-7H2,1H3
SMILES CCCCCCCC=O
IUPAC Name octanal
Molecular Weight (g/mol) 128.21
Formula Weight 128.1201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.