missing translation for 'onlineSavingsMsg'
Learn More

Octanal, 98%

Product Code. 11404273
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404273 1000 g 1000g
11414273 50 g 50g
11424273 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404273 Supplier Thermo Scientific Alfa Aesar Supplier No. A10901.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-13-0 | C8H16O | 128.22 g/mol

Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Air and moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-13-0
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00007029
InChI Key NUJGJRNETVAIRJ-UHFFFAOYSA-N
Synonym caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal
PubChem CID 454
ChEBI CHEBI:17935
IUPAC Name octanal
SMILES CCCCCCCC=O

Specifications

Melting Point 12°C to 15°C
Density 0.821
Boiling Point 169°C to 171°C
Flash Point 56°C (133°F)
Linear Formula CH3(CH2)6CHO
Refractive Index 1.4195
Quantity 1000 g
UN Number UN1191
Sensitivity Air Sensitive
Merck Index 14,1766
Solubility Information Slightly soluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material Octanal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.