missing translation for 'onlineSavingsMsg'
Learn More

Octamethyltrisiloxane, TRC

Product Code. 30425860
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425860 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425860 Supplier Toronto Research Chemicals Supplier No. TRCO2371505G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octamethyltrisiloxane
CAS 107-51-7
Molecular Formula C8 H24 O2 Si3
Quantity 5 g
Synonym Trisiloxane','1,1,1,3,3,5,5,5-octamethyl- (ACI), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane (ACI), Trisiloxane','octamethyl- (6CI','8CI','9CI), Dimethylbis(trimethylsiloxy)silane, Dow Corning OS 20, Dowsil OS 20, FRD 20, KF 96A1cs, KF 96L1.0, MDM, MDM (siloxane), Octamethyltrisiloxane, OS 20, OS 20 Fluid, Pentamethyl(trimethylsiloxy)disiloxane, Pentamethyl(trimethylsilyloxy)disiloxane, R 030007, Trisiloxane','1,1,1,3,3,5,5,5-octamethyl- (ACI), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane (ACI), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane, Trisiloxane','octamethyl- (6CI','8CI','9CI), Dimethylbis(trimethylsiloxy)silane, Dow Corning OS 20, Dowsil OS 20, FRD 20, KF 96A1cs, KF 96L1.0, MDM, MDM (siloxane), OS 20, OS 20 Fluid, Pentamethyl(trimethylsiloxy)disiloxane, Pentamethyl(trimethylsilyloxy)disiloxane, R 030007
InChI Formula InChI=1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
IUPAC Name dimethyl-bis(trimethylsilyloxy)silane
Molecular Weight (g/mol) 236.532
Formula Weight 236.108
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.