missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasiloxane, 98%

Product Code. 10286600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286600 100 g Glass bottle 100g
10226560 25 g Glass bottle 25g
10434945 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286600 Supplier Thermo Scientific Acros Supplier No. 216471000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-67-2 | C8H24O4Si4 | 296.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-67-2
Molecular Formula C8H24O4Si4
Molecular Weight (g/mol) 296.61
MDL Number MFCD00003269
InChI Key HMMGMWAXVFQUOA-UHFFFAOYSA-N
Synonym octamethylcyclotetrasiloxane, cyclotetrasiloxane, octamethyl, oktamethylcyklotetrasiloxan, cyclic dimethylsiloxane tetramer, omcts, nuc silicone vs 7207, union carbide 7207, silicone sf 1173, oktamethylzyklotetrasiloxan, unii-cz227117je
PubChem CID 11169
ChEBI CHEBI:25640
IUPAC Name 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C)(C)C)(C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 17.0°C to 18.0°C
Color Colorless
Density 0.9560g/mL
Boiling Point 175.0°C to 176.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3950 to 1.3970
Quantity 100 g
Beilstein 04, III, 1885
Merck Index 15, 6836
Specific Gravity 0.956
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in carbon tetrachloride
Formula Weight 296.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octamethylcyclotetrasiloxane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361f-Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.